2,3-Naphthalenedicarboxaldehyde, 95%
2,3-Naphthalenedicarboxaldehyde, 95%
2,3-Naphthalenedicarboxaldehyde, 95%
Thermo Scientific Chemicals

2,3-Naphthalenedicarboxaldehyde, 95%

CAS: 7149-49-7 | C12H8O2 | 184.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
View all
Glass bottle
Catalog number 294725000
Price (USD)
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7149-49-7
IUPAC Namenaphthalene-2,3-dicarbaldehyde
Molecular FormulaC12H8O2
InChI KeyZIPLKLQPLOWLTM-UHFFFAOYSA-N
SMILESO=CC1=C(C=O)C=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only