Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes
Thermo Scientific™

Nalgene™ Rapid-Flow™ Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes

Achieve fast flow rates and high throughput for increased filtration efficiency with Nalgene Rapid-Flow sterile disposable bottle top filters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Material (Membrane)Pore SizeFits Bottle Neck Size (Metric)
295-4545500 mLPES0.45 μm45mm
596-3320150 mLPES0.2 μm33mm
596-4520150 mLPES0.2 μm45mm
296-3345150 mLPES0.45 μm33mm
296-4545150 mLPES0.45 μm45mm
595-3320500 mLPES0.2 μm33mm
595-4520500 mLPES0.2 μm45mm
295-3345500 mLPES0.45 μm33mm
597-33201000 mLPES0.2 μm33mm
597-45201000 mLPES0.2 μm45mm
290-3320150 mLSFCA0.2 μm33mm
290-4520150 mLSFCA0.2 μm45mm
290-3345150 mLSFCA0.45 μm33mm
290-4545150 mLSFCA0.45 μm45mm
291-3320500 mLSFCA0.2 μm33mm
291-4520500 mLSFCA0.2 μm45mm
291-3345500 mLSFCA0.45 μm33mm
291-4545500 mLSFCA0.45 μm45mm
292-33201000 mLSFCA0.2 μm33mm
292-45201000 mLSFCA0.2 μm45mm
Catalog number 295-4545
Price (USD)
125.25
Online exclusive
167.00
Save 41.75
Case of 12
Volume (Metric):
500 mL
Material (Membrane):
PES
Pore Size:
0.45 μm
Fits Bottle Neck Size (Metric):
45mm
Price (USD)
125.25
Online exclusive
167.00
Save 41.75
Case of 12

Increase filtration efficiency. Thermo Scientific™ Nalgene™ Rapid-Flow™ single use bottle tops feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput. The last line of defense against cell culture contamination—Nalgene Rapid-Flow bottle tops are the best choice for the filtration of cell culture media, serum, additives and buffers.

  • Available in PES and SFCA membranes. Devices are color coded according to membrane.
  • Devices feature the fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse embryonic assay (MEA) tested
  • Non-pyrogenic and endotoxin free - Rapid-Flow filters meet the most rigorous testing standards for any vacuum filtration unit
  • Available with Fit for Purpose Sterile documentation
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Diameter (Metric)75 mm
Diameter (Metric) Membrane75 mm
Fits Bottle Neck Size (Metric)45mm
No. per Case12
No. per Pack12
TypeBottle Top Filter
Volume (Metric)500 mL
DisposableSingle-use
Filter TypeMembrane Filter
Fit for PurposeSterile
MaterialPolystyrene
Material (Membrane)PES
Pore Size0.45 μm
SterilitySterile
Unit SizeCase of 12

Frequently asked questions (FAQs)

Can I remove the filter membranes from the Nalgene Rapid-Flow Sterile Disposable Bottle Top Filters with PES, CN, SFCA or Nylon Membranes?

No, the filter membranes for the bottle top filters are molded in and cannot be removed.