2,6-Diaminopurine, 98%
2,6-Diaminopurine, 98%
2,6-Diaminopurine, 98%
Thermo Scientific Chemicals

2,6-Diaminopurine, 98%

CAS: 1904-98-9 | C5H6N6 | 150.14 g/mol
Have Questions?
Catalog NumberQuantityPackaging
2952700505 gGlass bottle
Catalog number 295270050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1904-98-9
IUPAC Name7H-purine-2,6-diamine; sulfuric acid
Molecular FormulaC5H8N6O4S
InChI KeyKYYLASPNTMEZTB-UHFFFAOYSA-N
SMILESOS(O)(=O)=O.NC1=NC(N)=C2NC=NC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow to light orange
Appearance (Form)Crystalline powder or granules
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only