4,6-Dimethyl-2-pyrone, 98%
4,6-Dimethyl-2-pyrone, 98%
4,6-Dimethyl-2-pyrone, 98%
Thermo Scientific Chemicals

4,6-Dimethyl-2-pyrone, 98%

CAS: 675-09-2 | C7H8O2 | 124.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 295350100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS675-09-2
IUPAC Name4,6-dimethyl-2H-pyran-2-one
Molecular FormulaC7H8O2
InChI KeyIXYLIUKQQQXXON-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow
Appearance (Form)Low melting crystalline mass
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only