Phosphoric acid, extra pure, 85% solution in water
Phosphoric acid, extra pure, 85% solution in water
Phosphoric acid, extra pure, 85% solution in water
Thermo Scientific Chemicals

Phosphoric acid, extra pure, 85% solution in water

CAS: 7664-38-2 | H3O4P | 97.994 g/mol
数量:
1 L
2.5 L
10 L
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 295700100
価格(JPY)
-
数量:
10 L
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7664-38-2
IUPAC Namephosphoric acid
Molecular FormulaH3O4P
InChI KeyNBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILESOP(O)(O)=O
さらに表示
Color scale=<35 APHA
Heavy metals (as Pb)=<10 ppm
Arsenic (As)=<2 ppm
Chloride (Cl)=<10 ppm
Titration with NaOH84.0 to 87.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only