Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru
Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru
Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru
Thermo Scientific Chemicals

Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru

CAS: 12289-94-0 | C16H26Ru | 319.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 295782500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12289-94-0
IUPAC NameBis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)
Molecular FormulaC16H26Ru
SMILES[Ru++].Cc(:c):c.Cc(:c):c.C1C\C=C/CC\C=C/1
Molecular Weight (g/mol)319.45
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Beige to brown
Melting point80°C to 84°C
Infrared spectrumConforms
NMR>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only