Hexamethylditin, 99%
Hexamethylditin, 99%
Hexamethylditin, 99%
Thermo Scientific Chemicals

Hexamethylditin, 99%

CAS: 661-69-8 | C6H18Sn2 | 327.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 295850010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS661-69-8
IUPAC Namehexamethyldistannane
Molecular FormulaC6H18Sn2
InChI KeyCCRMAATUKBYMPA-UHFFFAOYSA-N
SMILESC[Sn](C)(C)[Sn](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy colorless to white
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid or solid
Refractive index1.5370 to 1.5410 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only