3-Octylthiophene, 97%, Thermo Scientific Chemicals
3-Octylthiophene, 97%, Thermo Scientific Chemicals
3-Octylthiophene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Octylthiophene, 97%, Thermo Scientific Chemicals

CAS: 65016-62-8 | Molecular Formula: C12H20S | Molecular Weight: 196.36 g/mol
Have Questions?
Catalog NumberQuantityPackaging
29614010010 gGlass bottle
Catalog number 296140100
Price (USD)
149.00
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
149.00
Each
Chemical Identifiers
CAS65016-62-8
IUPAC Name3-octylthiophene
Molecular FormulaC12H20S
InChI KeyWQYWXQCOYRZFAV-UHFFFAOYSA-N
SMILESCCCCCCCCC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4900 to 1.4950 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only