(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%
(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%
Thermo Scientific Chemicals

(S)-(+)-2,3,7,7a-Tetrahydro-7a-methyl-1H-indene-1,5(6H)-dione, 90%

CAS: 17553-86-5 | C10H12O2 | 164.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 296470010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS17553-86-5
IUPAC Name(7aS)-7a-methyl-2,3,5,6,7,7a-hexahydro-1H-indene-1,5-dione
Molecular FormulaC10H12O2
InChI KeyFNYAZSZTENLTRT-JTQLQIEISA-N
SMILESC[C@]12CCC(=O)C=C1CCC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point55°C to 64°C
GC>=88 %
Infrared spectrumConforms
Specific optical rotation+330° to +360° (20°C, 589 nm) (c=1, toluene)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only