2-Bromophenylacetic acid, 98%
2-Bromophenylacetic acid, 98%
2-Bromophenylacetic acid, 98%
Thermo Scientific Chemicals

2-Bromophenylacetic acid, 98%

CAS: 18698-97-0 | C8H7BrO2 | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
Packaging:
Glass bottle
Catalog number 296650100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18698-97-0
IUPAC Name2-(2-bromophenyl)acetate
Molecular FormulaC8H6BrO2
InChI KeyDWXSYDKEWORWBT-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Melting point105°C to 110°C
Titration with NaOH>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only