Phenylhydrazine, 97%
Phenylhydrazine, 97%
Phenylhydrazine, 97%
Thermo Scientific Chemicals

Phenylhydrazine, 97%

CAS: 100-63-0 | C6H8N2 | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 296680025
Price (USD)
318.00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
318.00
Each
Chemical Identifiers
CAS100-63-0
IUPAC Namephenylhydrazine
Molecular FormulaC6H8N2
InChI KeyHKOOXMFOFWEVGF-UHFFFAOYSA-N
SMILESNNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light orange
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only