Azetidine, 98%
Azetidine, 98%
Azetidine, 98%
Thermo Scientific Chemicals

Azetidine, 98%

CAS: 503-29-7 | C3H7N | 57.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 296730050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS503-29-7
IUPAC Nameazetidine
Molecular FormulaC3H7N
InChI KeyHONIICLYMWZJFZ-UHFFFAOYSA-N
SMILESC1CNC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4290 to 1.4330 (20°C, 589 nm)
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only