Trimethylboroxine, 3.5M (50 wt.%) solution in THF
Trimethylboroxine, 3.5M (50 wt.%) solution in THF
Trimethylboroxine, 3.5M (50 wt.%) solution in THF
Thermo Scientific Chemicals

Trimethylboroxine, 3.5M (50 wt.%) solution in THF

CAS: 823-96-1 | C3H9B3O3 | 125.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 297520250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS823-96-1
IUPAC Nametrimethyl-1,3,5,2,4,6-trioxatriborinane
Molecular FormulaC3H9B3O3
InChI KeyGBBSAMQTQCPOBF-UHFFFAOYSA-N
SMILESCB1OB(C)OB(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to yellow
Assay40.0 to 60.0 % (ex Boron)
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only