L(-)-Lactic acid, lithium salt, 99%, pure
L(-)-Lactic acid, lithium salt, 99%, pure
L(-)-Lactic acid, lithium salt, 99%, pure
L(-)-Lactic acid, lithium salt, 99%, pure
Thermo Scientific Chemicals

L(-)-Lactic acid, lithium salt, 99%, pure

CAS: 27848-80-2 | C3H5O3Li | 96.01 g/mol
数量:
25 g
100 g
500 g
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 297611000
価格(JPY)
-
見積もりを依頼する
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS27848-80-2
IUPAC Namelithium(1+) 2-hydroxypropanoate
Molecular FormulaC3H5LiO3
InChI KeyGKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES[Li+].CC(O)C([O-])=O
さらに表示
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
Specific optical rotation-13° to -16° (20°C, 589 nm) (c=1, H2O)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only