2-(2-Bromoethyl)-1,3-dioxane, 98%
2-(2-Bromoethyl)-1,3-dioxane, 98%
2-(2-Bromoethyl)-1,3-dioxane, 98%
Thermo Scientific Chemicals

2-(2-Bromoethyl)-1,3-dioxane, 98%

CAS: 33884-43-4 | C6H11BrO2 | 195.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
29793010010 gGlass bottle
Catalog number 297930100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS33884-43-4
IUPAC Name2-(1-bromoethyl)-1,3-dioxane
Molecular FormulaC6H11BrO2
InChI KeyKXMZOKKPQZRPRN-UHFFFAOYNA-N
SMILESCC(Br)C1OCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.4800 to 1.4840 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only