1,3-Dioxolane, 99.5+%, pure, stabilized
1,3-Dioxolane, 99.5+%, pure, stabilized
1,3-Dioxolane, 99.5+%, pure, stabilized
Thermo Scientific Chemicals

1,3-Dioxolane, 99.5+%, pure, stabilized

CAS: 646-06-0 | C3H6O2 | 74.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 299480025
Price (USD)
216.65
Special offer
Online exclusive
Ends: 28-Dec-2026
255.00
Save 38.35
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
216.65
Special offer
Online exclusive
Ends: 28-Dec-2026
255.00
Save 38.35
Each
Chemical Identifiers
CAS646-06-0
IUPAC Name1,3-dioxolane
Molecular FormulaC3H6O2
InChI KeyWNXJIVFYUVYPPR-UHFFFAOYSA-N
SMILESC1COCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Refractive index1.3990 to 1.4010 (20°C, 589 nm)
Stabilizer60 to 100 ppm BHT
GC>=99.50 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only