Nalgene™ Reusable Filter Units
Thermo Scientific™
Nalgene™ Reusable Filter Units
Eliminate the need for a manifold or breakable glass vacuum flask. Nalgene Reusable Filter Holders with Receiver are complete, environmentally-friendly systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric) ReceiverUnit Size
300-41001000 mLCase of 4
300-4000250 mLCase of 4
300-4000PK
also known as 300-4000
250 mLEach
300-4050500 mLCase of 4
300-4050PK500 mLEach
300-4100PK1000 mLEach
Catalog number 300-4100
Price (USD)
1,261.80
Online exclusive
1,472.00
Save 210.20
Case of 4
Volume (Metric) Receiver:
1000 mL
Unit Size:
Case of 4
Price (USD)
1,261.80
Online exclusive
1,472.00
Save 210.20
Case of 4
Reusable filter units include two membrane support plates: a clear one-piece plate for sterilization applications and a two-piece white plate for analytical applications. Accepts 47 or 50 mm diameter membranes. Two side-arms allow connection to vacuum line. Tubing adapters accept 1/4- to 5/16-inch (6 to 8 mm) I.D. vacuum tubing.
Features
  • Replaces metal or breakable glass – major components are made of durable, transparent, autoclavable polysulfone
  • Use for sterilization or analysis. Filter holders come with sterilization and analytical membrane support plates
  • Designed to be used with vacuum or pressure
  • Graduated chambers for easy reference
Specifications
AutoclavableAutoclavable
No. per Pack1
Volume (Metric) Receiver1000 mL
Volume (Metric) Upper500 mL
MaterialPolysulfone
Unit SizeCase of 4