Nalgene™ PETG Biotainer™ Bottles
Thermo Scientific™

Nalgene™ PETG Biotainer™ Bottles

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
3005-42500 mL14 x 5 Bottles
3025-42125 mL20 x 5 Bottles
3005-70500 mL70 Bottles
3110-421 L7 x 5 Bottles
3110-351 L35 Bottles
3230-422 L4 x 5 Bottles
3230-202 L20 Bottles
3415-425 L6 Bottles
3415-165 L6 Bottles
Catalog number 3005-42
Price (USD)
738.00
Case of 70
Capacity (Metric):
500 mL
Quantity:
14 x 5 Bottles
Price (USD)
738.00
Case of 70

Eliminate the cost of cleaning, sterilizing and associated validations compared to bottles designed for reuse with Thermo Scientific™ Nalgene™ PETG Biotainer™ Bottles, ideal for prolonged storage of pH sensitive materials such as culture media. Certified sterile and ready-to-use, all Biotainer bottles are gamma irradiated to a Sterility Assurance Level (SAL) of 10–6.

  • Durable, crystal clear PETG for sample integrity
  • Closures with polyethylene foam core liners for leakproof† assurance
  • Custom options available for fluid transfer applications
  • Supported by extensive validation documentation
  • Suitable for storage in conditions from -40° to +70°C

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableNot Autoclavable
Capacity (English)16.9 fl. oz.
Certifications/ComplianceUSP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315
Closure Size38 mm
Diameter (English)3 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric)77 mm
Diameter (Metric) Inner Neck28 mm
DisposableSingle-use
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​
Height (English)6.93 in.
Height (Metric)176 mm
No. per Pack5/Pk.
Product TypeSquare Bottle
Quantity14 x 5 Bottles
ShapeSquare
Temperature Range (Metric)-70°C to 70°C
Workflow StepUpstream process solution storage. Upstream Validation
Capacity (Metric)500 mL
Closure IncludedYes
ColorTranslucent Blue
GraduatedYes
MaterialPET
MouthNarrow
SterilitySterile
Unit SizeCase of 70