Ethanol-d6, for NMR, anhydrous, 99 atom % D
Ethanol-d6, for NMR, anhydrous, 99 atom % D
Ethanol-d6, for NMR, anhydrous, 99 atom % D
Thermo Scientific Chemicals

Ethanol-d6, for NMR, anhydrous, 99 atom % D

CAS: 1516-08-1 | C2H6O | 52.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mL
5 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 300680050
Price (USD)
-
Quantity:
5 mL
Request bulk or custom format
Chemical Identifiers
CAS1516-08-1
IUPAC Name(²H₅)ethan-1-(²H)ol
Molecular FormulaC2H6O
InChI KeyLFQSCWFLJHTTHZ-LIDOUZCJSA-N
SMILES[2H]OC([2H])([2H])C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
D-Enrichment>=98.5 %
Appearance (Form)Liquid
Water=<1 % (Coulometric)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only