1-Octene, 99+%
1-Octene, 99+%
1-Octene, 99+%
Thermo Scientific Chemicals

1-Octene, 99+%

CAS: 111-66-0 | C8H16 | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Metal Drum
Catalog number 301250100
Price (USD)
340.65
Special offer
Online exclusive
Ends: 28-Dec-2026
400.00
Save 59.35
Each
Quantity:
10 L
Packaging:
Metal Drum
Request bulk or custom format
Price (USD)
340.65
Special offer
Online exclusive
Ends: 28-Dec-2026
400.00
Save 59.35
Each
Chemical Identifiers
CAS111-66-0
IUPAC Nameoct-1-ene
Molecular FormulaC8H16
InChI KeyKWKAKUADMBZCLK-UHFFFAOYSA-N
SMILESCCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.005 %
Refractive index1.4080 to 1.4100 (20°C, 589 nm)
Appearance (Form)Clear liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only