L-Selenocystine, 95%, Thermo Scientific Chemicals
L-Selenocystine, 95%, Thermo Scientific Chemicals
L-Selenocystine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Selenocystine, 95%, Thermo Scientific Chemicals

CAS: 29621-88-3 | C6H12N2O4Se2 | 334.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 301860010
Price (USD)
235.65
Online exclusive
262.00
Save 26.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
235.65
Online exclusive
262.00
Save 26.35
Each
Chemical Identifiers
CAS29621-88-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange to brown
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration excess NaOH94.0 to 106.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only