Hypersil™ MOS C8 HPLC Columns
Thermo Scientific™

Hypersil™ MOS C8 HPLC Columns

Use as a general-purpose C8 phase for acidic and neutral analytes.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Particle Size
30210-204630200 mm4.6 mm10 μm
30205-05463050 mm4.6 mm5 μm
30205-104630100 mm4.6 mm5 μm
30205-124630125 mm4.6 mm5 μm
30205-154630150 mm4.6 mm5 μm
30205-154030150 mm4 mm5 μm
30205-254030250 mm4 mm5 μm
30205-202130200 mm2.1 mm5 μm
30205-254630250 mm4.6 mm5 μm
30205-204630200 mm4.6 mm5 μm
30205-01400110 mm4 mm5 μm
30205-253030250 mm3 mm5 μm
30210-254630250 mm4.6 mm10 μm
Catalog number 30210-204630
Price (USD)
-
Length (Metric):
200 mm
Diameter (Metric):
4.6 mm
Particle Size:
10 μm

For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.

Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

  • Reliable, less retentive phase
  • Long column lifetimes, even under basic conditions
  • Manufactured under ISO 9001-2000 quality guidelines
  • Affordable

Ordering Information:

Other column dimensions are available. Also available in 10 μm. Please call Customer Service for more information.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Column FormatAnalytical Column
Column TypeReversed Phase
EndcappedNo
For Use WithHPLC
Max. Pressure5800 psi (400 bar)
Packing MaterialSpherical, Fully Porous Type A Silica
Stationary PhaseC8
Surface Area170 m²/g
Temperature60°C
USP TypeL7
Carbon Load6.5%
Diameter (Metric)4.6 mm
Length (Metric)200 mm
Particle Size10 μm
PhaseReversed Phase
Pore Size120 Å
Product LineHypersil Classical
pH2 to 8
Unit SizeEach