4,6-Dimethyldibenzothiophene, 95%
4,6-Dimethyldibenzothiophene, 95%
4,6-Dimethyldibenzothiophene, 95%
Thermo Scientific Chemicals

4,6-Dimethyldibenzothiophene, 95%

CAS: 1207-12-1 | C14H12S | 212.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 302190010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1207-12-1
IUPAC Name3,12-dimethyl-8-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
Molecular FormulaC14H12S
InChI KeyKMPJENUWHPZRGZ-UHFFFAOYSA-N
SMILESCC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=94 %
Infrared spectrumConforms
Appearance (Color)Light yellow to beige to light brown
Melting point150°C to 156°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only