Product Image
Thermo Scientific™
Guardcap H Vial Cap, Pack of 125
Automate inline removal of cations, metals, and neutralization of bases without extra labor, time, and complexity using Thermo Scientific™ Dionex™ Guardcap™ H vial caps.
Have Questions?
Catalog NumberFor Use With (Equipment)
3025045 mL AS-DV PolyVials
Catalog number 302504
Price (USD)
497.00
Each
For Use With (Equipment):
5 mL AS-DV PolyVials
Price (USD)
497.00
Each
Remove high levels of alkaline earth and transition metals, neutralize basic samples, and filter particulates with the Thermo Scientific™ Dionex™ Guardcap™ H vial caps. Designed for use with Thermo Scientific™ Dionex™ AS-DV Autosampler PolyVials, Dionex Guardcap H vial caps contain sulfonated strong cation exchange resin in the hydronium form that is embedded in a high density polyethylene matrix. Dionex Guardcap H vial caps seamlessly integrate automated inline sample preparation into your existing sample workflow without the need for additional labor, time, and complexity.
Each Guardcap H vial cap:
  • Removes 1000mg/L divalent cation (e.g., calcium or transition metals) from 800μL of sample
  • Neutralizes up to 500μL of 50mM NaOH or other base
  • Filters more than 80% of particulate matter greater than 20μm in diameter
Obtain Superior Analytical Results with Greatly Reduced Effort
Increase the lifetime of your analytical columns and suppressors by removing metals, matrix ions and particulates that can result in fouling.
Convenient and Easy to Use
Dionex Guardcap H vial caps come in the familiar filter cap format used with Dionex PolyVials. If you can load a filter cap on a Dionex PolyVial, you can use Dionex Guardcap H for automated inline sample preparation. Unlike Dionex OnGuard™ II H cartridges, Dionex Guardcap H vial caps require no pretreatment procedures prior to use.
Specifications
DescriptionGuardcap H Vial Cap, Pk. of 125
For Use With (Equipment)5 mL AS-DV PolyVials
MaterialOnGuard H Resin
QuantityEach 1
Product TypeFunctionalized Vial Cap
Unit SizeEach