Dionex™ Guardcap™ HRP Vial Caps
Dionex™ Guardcap™ HRP Vial Caps
Actual product may vary
Dionex™ Guardcap™ HRP Vial Caps
Dionex™ Guardcap™ HRP Vial Caps
Dionex™ Guardcap™ HRP Vial Caps
Dionex™ Guardcap™ HRP Vial Caps
Dionex™ Guardcap™ HRP Vial Caps
Remove hydrophobic organics and surfactants from IC samples without additional labor, time, or complexity using these Dionex Guardcap HRP vial caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
302798Functionalized Vial Cap with Transfer Line
302799Functionalized Vial Cap
Catalog number 302798
Price (USD)
89.20
Each
Add to cart
Type:
Functionalized Vial Cap with Transfer Line
Price (USD)
89.20
Each
Add to cart
Remove particulates and hydrophobic organic substances including non-ionic surfactants, hydrocarbons, aromatic dyes, and some carboxylic acids with Thermo Scientific™ Dionex™ Guardcap™ HRP Vial Caps. Designed for ion chromatography analyses using Thermo Scientific Dionex AS-DV autosampler PolyVials, Dionex Guardcap HRP vial caps contain hydrophilic reversed-phase resin embedded in a high density polyethylene filter matrix. Dionex Guardcap HRP vial caps seamlessly bring the power of automated inline sample preparation to your existing sample workflow without adding more labor, time, or complexity.
Each Guardcap HRP vial cap:
  • Arrives pre-wetted and ready for immediate use
  • Saves time and effort vs. traditional offline sample preparation cartridges
  • Protects columns and suppressors
  • Removes up to 0.01% non-ionic surfactant (e.g., Triton™ x-405) from 500μL of sample
  • Removes up to 20mg protonated carboxylic acids (e.g., benzoic acid at pH <4)
  • Leaves sample pH unchanged
  • Filters better than 80% of particulate matter greater than 20μm in diameter
Obtain superior analytical results with greatly reduced effort
Increase the lifetime of your analytical columns and suppressors by removing particulates and hydrophobic organic substances including non-ionic surfactants, hydrocarbons, aromatic dyes, and some carboxylic acids that can foul them using automated inline sample preparation. Sample preparation is performed inline, within the Dionex Guardcap vial cap, and automated by the autosampler. Dionex Guardcap vial caps can improve the reproducibility of analyses because important parameters such as timing, flow rate, and delivery volume are easily set and controlled by the autosampler.
Convenient and Easy to Use
Dionex Guardcap HRP vial caps come in the familiar filter cap format used with Dionex PolyVials. Unlike traditional offline sample preparation cartridges, Dionex Guardcap HRP vial caps require no time-consuming pretreatment procedures prior to use--simply install these functionalized vial caps on your sample vials and load your autosampler.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionDionex Guardcap HRP Vial Caps, Pk. of 12
For Use With (Equipment)5 mL AS-DV PolyVials
Includes70 cm reduced volume sample transfer line
MaterialHydrophilic Reversed Phase Resin
QuantityEach
TypeFunctionalized Vial Cap with Transfer Line
Unit SizeEach