Scopoletin, 95%
Scopoletin, 95%
Scopoletin, 95%
Thermo Scientific Chemicals

Scopoletin, 95%

CAS: 92-61-5 | C10H8O4 | 192.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Catalog number 302900500
Price (USD)
97.00
Each
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
97.00
Each
Chemical Identifiers
CAS92-61-5
IUPAC Name7-hydroxy-6-methoxy-2H-chromen-2-one
Molecular FormulaC10H8O4
InChI KeyRODXRVNMMDRFIK-UHFFFAOYSA-N
SMILESCOC1=C(O)C=C2OC(=O)C=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point200°C to 207°C
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=94.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only