ProPac™ Elite WCX HPLC Column

ProPac™ Elite WCX HPLC Column

Green features

This column provides consistent separations of therapeutic proteins including monoclonal antibodies, antibody drug conjugates, biosimilars and charge variants.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Quantity
302976150 mm4 mm3 Columns/1 Lot
30302850 mm2 mm1 Column
303027150 mm2 mm1 Column
303026250 mm2 mm1 Column
30297350 mm4 mm1 Column
302972150 mm4 mm1 Column
303025250 mm4 mm1 Column
302977150 mm4 mm3 Columns/3 Lots
303061250 mm4 mm3 Columns/1 Lot
303062250 mm4 mm3 Columns/3 Lots
303355150 mm9 mm1 Column
Catalog number 302976
Price (USD)
5,440.98
Pack of 3
Length (Metric):
150 mm
Diameter (Metric):
4 mm
Quantity:
3 Columns/1 Lot
Price (USD)
5,440.98
Pack of 3

Discover the unique Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column. Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants.

The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.

Product Highlights
  • Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
  • High efficiency with reproducible separa-tions
  • High recovery with low carry-over
  • Wide pH operating range: 2 – 12
  • High temperature stability: up to 60°C
  • High throughput
  • Compatible with CX-1 pH Gradient Buffers
Specifications
Column FormatAnalytical Column
Column TypeIon Exchange
Flow Rate0.4 to 1.0 mL/min
Green FeaturesFSC
Max. Pressure4500 psi (310 bar)
Mobile Phase CompatibilityRecommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O
Packing MaterialNonporous DVB Particles with Hydrophilic Coating
Quantity3 Columns/1 Lot
Stationary PhaseCarboxylate
Temperature10°C to 60°C
Diameter (Metric)4 mm
Length (Metric)150 mm
Particle Size5 μm
PhaseIon Exchange
Pore SizeNonporous
Product LineProPac
TechniqueBio-LC
pH2 to 12
Unit SizePack of 3