Dionex™ Guardcap™ AEX Vial Caps
Thermo Scientific™
Dionex™ Guardcap™ AEX Vial Caps
Remove anions from IC samples and neutralize acidic samples without additional labor, time, and complexity using Thermo Scientific™ Dionex™ Guardcap™ AEX vial caps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
302982Functionalized Vial Cap
302981Functionalized Vial Cap with Transfer Line
Catalog number 302982
Price (USD)
438.00
Each
Product Type:
Functionalized Vial Cap
Price (USD)
438.00
Each
Remove high levels of anions, neutralize acidic samples, and filter particulates with Thermo Scientific™ Dionex™ Guardcap™ AEX vial caps. Designed for use with Thermo Scientific Dionex AS-DV autosampler PolyVials, Dionex Guardcap AEX vial caps contain strong anion-exchange resin in the borate form embedded in a high density polyethylene matrix. Dionex Guardcap AEX vial caps seamlessly bring the power of automated inline sample preparation to your existing sample workflow without the need for additional labor, time, and complexity.
Each Guardcap AEX vial cap:
  • Saves time and effort vs. traditional offline sample preparation cartridges.
  • Removes up to 800 mg/L anions (e.g., sulfate) from 250 μL of sample.
  • Neutralizes up to 250 μL of 30 mM MSA, HCl, or other monovalent acid.
  • Filters better than 80% of particulate matter greater than 20 μm in diameter.
Obtain Superior Analytical Results with Greatly Reduced Effort
Increase the lifetime of your analytical columns and suppressors by removing anions and particulates that can foul them. Sample preparation is performed inline, within the Dionex Guardcap vial cap, and automated by the autosampler. Dionex Guardcap vial caps can improve the reproducibility of analyses because important parameters such as timing, flow rate, and delivery volume are easily set and controlled by the autosampler.
Convenient and Easy-to-Use
Dionex Guardcap AEX vial caps come in the familiar filter cap format used with Dionex PolyVials. Unlike traditional offline sample preparation cartridges, Dionex Guardcap AEX vial caps require no time-consuming pretreatment procedures prior to use- simply install these functionalized vial caps on your sample vials and your samples are ready for analysis.
Specifications
For Use With (Equipment)5 mL AS-DV PolyVials
IncludesNo accessories
MaterialBorate form anion-exchange resin
QuantityEach
Product TypeFunctionalized Vial Cap
Unit SizeEach