4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
4-(Trifluoromethyl)benzoyl chloride, 97%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzoyl chloride, 97%

CAS: 329-15-7 | C8H4ClF3O | 208.56 g/mol
数量:
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 303420010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS329-15-7
IUPAC Name4-(trifluoromethyl)benzoyl chloride
Molecular FormulaC8H4ClF3O
InChI KeyOXZYBOLWRXENKT-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)C(Cl)=O
さらに表示
Appearance (Color)Clear colorless to yellow
Refractive index1.4755 to 1.4775 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only