Select Agar™, powder
Invitrogen™

Select Agar™, powder

Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilizedRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
303910492.5 kg
30391023500 g
Catalog number 30391049
Price (USD)
852.65
Online exclusive
888.00
Save 35.35
Each
Quantity:
2.5 kg
Price (USD)
852.65
Online exclusive
888.00
Save 35.35
Each

Agar is a mixture of polysaccharides extracted from red seaweed cell walls. It has unique gelling properties that are utilized in many industrial and laboratory applications. Solid culture medium is created by adding  agar to media broth (typical final concentration is 1–2%). Solid media cell culture allows the isolation of individual E. coli colonies and is fundamental in cloning workflows.

Agar is used in diagnostic laboratories to detect pathogens from contaminated food or water. Also, it can be used to prepare selective media for saccharomyces cerevisiae and other yeasts used in various molecular microbiology procedures (e.g., YPD Agar). In addition, agar has been used in tissue double embedding for histopathology, hydrogel creation, and tissue engineering applications.

Primary applications
• Making solid media to grow cells (e.g., bacteria, fungi, yeast)
• To isolate E. coli clones with plasmid on a selective media
• Tissue engineering, hydrogel production

Look for more media that fit your needs
We offer other ready-to-use media formulations, such as LB (Lennox), Terrific Broth or S.O.C recovery medium.
Explore other media formats for bacterial growth.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatBottle
Media TypeSelect Agar
Preparation MethodAutoclave
Quantity2.5 kg
Shipping ConditionRoom Temperature
Product TypeDehydrated Culture Media
SpeciesEscherichia coli, Saccharomyces cerevisiae
Target Organism ClassColiforms, Fungi
Unit SizeEach
Contents & Storage
• 1 x 2.5 kg bottle Select Agar Powder

Store at room temperature.