Sodium metatungstate hydrate, crystalline
Thermo Scientific Chemicals

Sodium metatungstate hydrate, crystalline

CAS: 652154-12-6 | H2Na6O40W12·xH2O | 2986.13 g/mol
数量:
50 g
250 g
1 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 304090500
価格(JPY)
-
数量:
50 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS652154-12-6
Appearance (Color)White to light yellow
Assay>=85 % (WO3)
Appearance (Form)Crystalline powder or beads
Stabilizer=<200 ppm (Cr-salt) Typical
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only