Cycloheptene, 95%, stabilized
Cycloheptene, 95%, stabilized
Cycloheptene, 95%, stabilized
Thermo Scientific Chemicals

Cycloheptene, 95%, stabilized

CAS: 628-92-2 | C7H12 | 96.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 304260050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS628-92-2
IUPAC Namecycloheptene
Molecular FormulaC7H12
InChI KeyZXIJMRYMVAMXQP-UHFFFAOYSA-N
SMILESC1CCC=CCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Stabilizer0.1 % BHT
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only