Potassium chloride, 99.0-105.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Potassium chloride, 99.0-105.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Catalog number 3045-01
Price (USD)
-
Quantity:
500 g
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorWhite
Melting Point773°C
PackagingPoly bottle, tamper-evident cap
Boiling Point1420°C (1013 hPa)
GradeMulti-Compendial/USP
Molecular Weight (g/mol)74.55
Percent Purity99 to 105%
Physical FormSolid
Quantity500 g
pH7 (15°C, saturated solution in water)
Unit SizeEach