Potassium chloride, 99.0-105.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™
Avantor™

Potassium chloride, 99.0-105.0% (dried basis), crystals, USP, Multi-Compendial, GMP, J.T.Baker™

This product is GMP Manufactured and meets USP, JP, BP and EP specifications.Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Catalog number 3045-09
Price (MXN)
-
Quantity:
100 lb.
This product is GMP Manufactured and meets USP, JP, BP and EP specifications.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
ColorWhite
Melting Point773°C
PackagingPolyethylene drum, open head
Boiling Point1420°C (1013 hPa)
GradeMulti-Compendial/USP
Molecular Weight (g/mol)74.55
Percent Purity99 to 105%
Physical FormSolid
Quantity100 lb.
pH7 (15°C, saturated solution in water)
Unit SizeEach