4-Fluoro-3-nitrobenzoic acid, 98%
4-Fluoro-3-nitrobenzoic acid, 98%
4-Fluoro-3-nitrobenzoic acid, 98%
Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzoic acid, 98%

CAS: 453-71-4 | C7H4FNO4 | 185.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 305060050
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.90
Save 6.25
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.90
Save 6.25
Each
Chemical Identifiers
CAS453-71-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point119°C to 126°C
Appearance (Color)Light yellow to tan
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only