2-Bromo-2-methylpropionic acid, 98%
2-Bromo-2-methylpropionic acid, 98%
2-Bromo-2-methylpropionic acid, 98%
2-Bromo-2-methylpropionic acid, 98%
Thermo Scientific Chemicals

2-Bromo-2-methylpropionic acid, 98%

CAS: 2052-01-9 | C4H7BrO2 | 167.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 305201000
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.30
Save 13.65 (15%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
76.65
Special offer
Online exclusive
Ends: 30-Jun-2026
90.30
Save 13.65 (15%)
Each
Chemical Identifiers
CAS2052-01-9
IUPAC Name2-bromo-2-methylpropanoic acid
Molecular FormulaC4H7BrO2
InChI KeyXXSPGBOGLXKMDU-UHFFFAOYSA-N
SMILESCC(C)(Br)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to orange
Titration with NaOH>=97.5 %
Melting point41°C to 46°C
Appearance (Form)Semitransparent crystalline powder or crystals or
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only