Sodium thiomethoxide, 95%, pure
Sodium thiomethoxide, 95%, pure
Sodium thiomethoxide, 95%, pure
Thermo Scientific Chemicals

Sodium thiomethoxide, 95%, pure

CAS: 5188-07-8 | CH3NaS | 70.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 305710010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5188-07-8
IUPAC Namesodium methylsulfanide
Molecular FormulaCH3NaS
InChI KeyRMBAVIFYHOYIFM-UHFFFAOYSA-M
SMILES[Na+].C[S-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granular powder
Appearance (Color)White to light beige
Titration Iodimetric94 to 106 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only