Methyl 4-(bromomethyl)benzoate, 98%
Methyl 4-(bromomethyl)benzoate, 98%
Methyl 4-(bromomethyl)benzoate, 98%
Thermo Scientific Chemicals

Methyl 4-(bromomethyl)benzoate, 98%

CAS: 2417-72-3 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 306100050
Price (USD)
46.65
Online exclusive
51.60
Save 4.95
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
46.65
Online exclusive
51.60
Save 4.95
Each
Chemical Identifiers
CAS2417-72-3
IUPAC Namemethyl 4-(bromomethyl)benzoate
Molecular FormulaC9H9BrO2
InChI KeyNLWBJPPMPLPZIE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to yellow
Melting point52°C to 57°C
GC>=97.5 %
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only