(Triphenylphosphoranylidene)acetaldehyde, 97%
(Triphenylphosphoranylidene)acetaldehyde, 97%
(Triphenylphosphoranylidene)acetaldehyde, 97%
Thermo Scientific Chemicals

(Triphenylphosphoranylidene)acetaldehyde, 97%

CAS: 2136-75-6 | C20H17OP | 304.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 307490250
Price (USD)
168.65
Special offer
Online exclusive
Ends: 28-Dec-2026
198.00
Save 29.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
168.65
Special offer
Online exclusive
Ends: 28-Dec-2026
198.00
Save 29.35
Each
Chemical Identifiers
CAS2136-75-6
IUPAC Name2-(triphenyl-λ⁵-phosphanylidene)acetaldehyde
Molecular FormulaC20H17OP
InChI KeyCQCAYWAIRTVXIY-UHFFFAOYSA-N
SMILESO=CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Appearance (Color)Cream to pink to orange to brown
Titration with HClO496.0 to 104.0 % (on dry substance)
Water=<4.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only