Phloretin, 98%
Phloretin, 98%
Phloretin, 98%
Thermo Scientific Chemicals

Phloretin, 98%

CAS: 60-82-2 | C15H14O5 | 274.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Catalog number 307651000
Price (USD)
-
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS60-82-2
IUPAC Name3-(4-hydroxyphenyl)-1-(2,4,6-trihydroxyphenyl)propan-1-one
Molecular FormulaC15H14O5
InChI KeyVGEREEWJJVICBM-UHFFFAOYSA-N
SMILESOC1=CC=C(CCC(=O)C2=C(O)C=C(O)C=C2O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only