3-Phenyltoluene, 95%
3-Phenyltoluene, 95%
3-Phenyltoluene, 95%
Thermo Scientific Chemicals

3-Phenyltoluene, 95%

CAS: 643-93-6 | C13H12 | 168.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 309840010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS643-93-6
IUPAC Name3-methyl-1,1'-biphenyl
Molecular FormulaC13H12
InChI KeyNPDIDUXTRAITDE-UHFFFAOYSA-N
SMILESCC1=CC=CC(=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=94 %
Appearance (Color)Clear light yellow
Refractive index1.6010 to 1.6050 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only