Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfide, 90+%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 310180500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1313-82-2
IUPAC Namedisodium sulfanediide
Molecular FormulaNa2S
InChI KeyGRVFOGOEDUUMBP-UHFFFAOYSA-N
SMILES[Na+].[Na+].[S--]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Appearance (Form)Powder or flakes and/or lumps
Titration Iodimetric>=90.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only