Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, 15mL - FAQs
Nalgene™ PPCO Conical-Bottom Centrifuge Tubes, 15mL - FAQs
View additional product information for Nalgene™ PPCO Conical-Bottom Centrifuge Tubes - FAQs (3103-0015PK, 3103-0050, 3103-0015, 3103-0050PK)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.