5-Hydroxyanthranilic acid, 98%
5-Hydroxyanthranilic acid, 98%
5-Hydroxyanthranilic acid, 98%
Thermo Scientific Chemicals

5-Hydroxyanthranilic acid, 98%

CAS: 394-31-0 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 310310100
Price (USD)
66.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.90
Save 11.25
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 28-Dec-2026
77.90
Save 11.25
Each
Chemical Identifiers
CAS394-31-0
IUPAC Name2-amino-5-hydroxybenzoic acid
Molecular FormulaC7H7NO3
InChI KeyHYNQTSZBTIOFKH-UHFFFAOYSA-N
SMILESNC1=CC=C(O)C=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystalline powder or crystals
Titration with NaOH97.5 to 102.5 %
Appearance (Color)White to grey-brown to brown
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only