Phenylmaleic anhydride, 99%
Phenylmaleic anhydride, 99%
Phenylmaleic anhydride, 99%
Thermo Scientific Chemicals

Phenylmaleic anhydride, 99%

CAS: 36122-35-7 | C10H6O3 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 310400010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36122-35-7
IUPAC Name3-phenyl-2,5-dihydrofuran-2,5-dione
Molecular FormulaC10H6O3
InChI KeyQZYCWJVSPFQUQC-UHFFFAOYSA-N
SMILESO=C1OC(=O)C(=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or needles
Appearance (Color)White to light brown
Infrared spectrumConforms
GC>=98.5 %
Melting point119°C to 123°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only