Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes, 50mL, 102mm - FAQs
Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes, 50mL, 102mm - FAQs
View additional product information for Nalgene™ High-Speed Round-Bottom PPCO Centrifuge Tubes - FAQs (3110-0500PK, 3110-9500, 3110-0120, 3110-1000, 3110-0150, 3110-1000PK, 3110-9500PK, 3110-0160, 3110-0380, 3110-0120PK, 3110-0500, 3110-0160PK, 3110-0150PK, 3110-0380PK)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.