Forma™ Series II 3110 Water-Jacketed CO2 Incubators - FAQs
View additional product information for Forma™ Series II Water-Jacketed CO2 Incubator, 184L - FAQs (3111, 3110, 3121, 3131, 3120, 3130, 3141, 3140)
1 product FAQs found
Do you supply an HPLC chromatogram for Amyloid beta 1-42?
No, we don't provide a chromatogram since our Amyloid beta 1-42 is supplied as a kit component. Note that Zagorski, et al. (1999) points out that Amyloid beta 1-42 elutes from HPLC columns as an extremely broad peak with poor resolution, and impurities which co-elute with the broad Amyloid beta 1-42 band might not be resolved into separate peaks. When Amyloid beta 1-42-containing fractions are collected from the HPLC column and then re-injected, multiple peaks are still observed. Both of these observations, the broad peak and the formation of multiple peaks upon re-injection, are consistent with aggregation of Amyloid beta 1-42 under these HPLC conditions. Another limitation is that Amyloid beta has been observed to adhere irreversibly to a variety of common HPLC columns. At this time, HPLC analysis is not applicable for assessing Amyloid beta 1-42 purity.
Reference:
Zagorski, M.G., J. Yang, H. Shao, K. Ma, H. Zeng, and A. Hong (1999) Methodological and chemical factors affecting amyloid? peptide amyloidogenicity. Meth. Enzymol. 309:189-204.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.