Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™
Thermo Scientific™

Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™

Conduct high-speed centrifugation of aggressive chemicals with these tubes, suited for chloroform and phenol extractions in refrigerated machines up to 50,000 x g.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric)Unit Size
3114-0050PK
also known as 3114-0050
50 mL28.8 mmPack of 2
3114-001010 mL16 mmCase of 10
3114-0010PK
also known as 3114-0010
10 mL16 mmPack of 2
3114-003030 mL25.7 mmCase of 10
3114-0030PK
also known as 3114-0030
30 mL25.7 mmPack of 2
3114-005050 mL28.8 mmCase of 10
Catalog number 3114-0050PK
also known as 3114-0050
Price (USD)
211.16
Online exclusive
234.63
Save 23.47
Pack of 2
Capacity (Metric):
50 mL
Diameter (Metric):
28.8 mm
Unit Size:
Pack of 2
Price (USD)
211.16
Online exclusive
234.63
Save 23.47
Pack of 2

Conduct high-speed centrifugation of aggressive chemicals with Thermo Scientific™ Nalgene™ Oak Ridge High-Speed Centrifuge Tubes made with Teflon™ fluoropolymer and Closure made with Tefzel™, specifically suited for chloroform and phenol extractions.

Features:
  • Withstand temperatures from -100° to +150°C (-148° to +302°F)
  • Leakproof† closures prevent material loss under ordinary use for most spins <10,000 × g
  • When equipped with a Nalgene sealing cap assemblies (sold separately), 30mL and 50mL tubes can be spun up to 50,000 x g
  • Must be filled to at least 100% total capacity for proper performance. 
  • Autoclavable/Biohazard/Leakproof† when used with supplied Nalgene closure /PTFE FEP

Ordering Information:

Optional sealing cap assemblies available — DS3131-0020 (for 30mL tube) and DS3131-0024 (for 50mL tube). Cat. No. 3114-0050 has a brim capacity of only 42mL but is designed to fit standard rotor.

Includes:

Screw caps. Additional sealing cap assembly available separately (DS3131-series).

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Closure MaterialETFE
Diameter (Metric)28.8 mm
No. per Case10
No. per Pack2
Product TypeCentrifuge Tube
ShapeRound
Writing AreaNo
AutoclavableAutoclavable
Bottom ShapeRound
Capacity (Metric)50 mL
Closure TypeScrew Cap
GraduatedNo
MaterialFEP
Max. RCF50,000 x g (with sealing cap assembly)
SterilityNon-sterile
Unit SizePack of 2