Nalgene™ Boston Round Narrow-Mouth HDPE Bottles with Closure: Bulk Pack, 1000mL - FAQs
Nalgene™ Boston Round Narrow-Mouth HDPE Bottles with Closure: Bulk Pack, 1000mL - FAQs
View additional product information for Nalgene™ Boston Round Narrow-Mouth HDPE Bottles with Closure: Bulk Pack - FAQs (312002-0004, 312002-0001, 312002-0002, 312002-0008, 312002-0016, 312002-0006, 312002-0032)
1 product FAQs found
What is the recommended torque for my bottle closure size for the Nalgene Boston Round Narrow-Mouth HDPE Bottles with Closure: Bulk Pack (Cat. No. 312002-xxxx)?
Please refer to the following chart for recommended torque based on your closure size.