Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack
Thermo Scientific™

Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure: Bulk Pack

Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals in these light-weight bottles which are leakproof when used with Nalgene closures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
312187-000130 mL28-415 mm
312187-000260 mL28-415 mm
312187-0004125 mL38-415 mm
312187-0008250 mL43-415 mm
312187-0016500 mL53-415 mm
312187-00321000 mL63-415 mm
Catalog number 312187-0001
Price (USD)
1,119.92
Online exclusive
1,244.35
Save 124.43
Case of 1000
Capacity (Metric):
30 mL
Closure Size:
28-415 mm
Price (USD)
1,119.92
Online exclusive
1,244.35
Save 124.43
Case of 1000

Ship and store diagnostics, biologicals, specialty chemicals, reagents, adhesives and veterinary pharmaceuticals with Thermo Scientific™ Nalgene™ Wide-Mouth PPCO Packaging Bottles with Closure. These durable, autoclavable bottles are designed for shipping and storage.

Features:
  • Wide mouth for easy filling and dispensing of powders
  • Injection blow molded for uniform wall thickness and strength
  • Made from high-purity, food-grade resins; no plasticizers, fillers or extenders
  • Polypropylene closures included
  • Leakproof† when used with Nalgene closures

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

A sample product certificate providing Cytotoxicity, USP Class VI, FDA Compliance—21 CFR and other regulatory compliance information is available or can be requested from our Certificates of Compliance Database

Specifications
AutoclavableAutoclavable
Capacity1 oz., 30 mL
Capacity (English)1 oz.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorNatural, translucent
Closure MaterialPolypropylene
Closure Size28-415 mm
Closure TypeScrew (Bulk Packed, Unassembled)
DescriptionWide-Mouth Packaging Bottles
Diameter (Metric)34 mm
Diameter (Metric) Inner Neck21 mm
Height (English)2.3 in. (With Closure); 2.2 in. (Without Closure)
Height (Metric)63 mm
Neck Finish28 mm
No. per Case1000
No. per Pack1000
Product TypePacker Bottle
ShapeRound
Width (English)1.35 in.
Capacity (Metric)30 mL
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialPPCO
MouthWide
Unit SizeCase of 1000

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.